accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)OH(R)NH2OHO

CAS RN 80-68-2

IUPAC Name: (2R,3R)-2-amino-3-hydroxybutanoic acid
Synonyms: allo-threonine|L-allo-threonine|D-allo-threonine|D-allothreonine|allothreonine, D-|(R)-allo-threonine|(R)-allothreonine|(2R,3R)-allothreonine
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: AYFVYJQAPQTCCC-PWNYCUMCSA-N
SMILES: C[C@@H](O)[C@@H](N)C(O)=O
Percent Purity 
  • (2)
  • (1)
  • (4)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (4)
  • (5)
  • (2)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (3)
  • (6)
  • (2)
  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (5)
  • (2)
  • (1)
  • (2)