Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Threonine, >99%, MP Biomedicals™

Catalog No. ICN10305091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10305091 1 kg
ICN10305010 10 g
ICN10305025 25 g
ICN10305080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10305091 Supplier MP Biomedicals, Inc Supplier No. 0210305091
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 80-68-2 | C4H9NO3 | 119.12 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-68-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00063722
InChI Key AYFVYJQAPQTCCC-PWNYCUMCSA-N
Synonym dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l
PubChem CID 205
ChEBI CHEBI:38263
SMILES C[C@@H](O)[C@@H](N)C(O)=O

Specifications

Color White
IUPAC Name 2-amino-3-hydroxybutanoic acid
Quantity 1 kg
Formula Weight 119.1
Percent Purity >99%
Physical Form Powder
Chemical Name or Material DL-Threonine
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.