Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Threonine (contains DL-Allothreonine) 97.0+%, TCI America™
SDP

Catalog No. T310525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T310525G 25 g
T3105100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T310525G Supplier TCI America Supplier No. T310525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 80-68-2 | C4H9NO3 | 119.12 g/mol

Chemical Identifiers

CAS 80-68-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00063722
InChI Key AYFVYJQAPQTCCC-PWNYCUMCSA-N
Synonym dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l
PubChem CID 205
ChEBI CHEBI:38263
IUPAC Name (2R,3R)-2-amino-3-hydroxybutanoic acid
SMILES C[C@@H](O)[C@@H](N)C(O)=O

Specifications

Melting Point 230°C
Color White
Quantity 25 g
Formula Weight 119.12
Percent Purity ≥97.0% (T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material DL-Threonine (contains DL-Allothreonine)
EINECSNumber (9)-1584
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.