Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLEC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104537 Supplier Invitrogen™ Supplier No. PA5104537
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DLEC1.

This gene contains 37 exons, spans approximately 59-kb, and is located in the 3p22-p21.3 chromosomal segment that is commonly deleted in various carcinomas. Several alternatively spliced transcripts have been observed that contain disrupted coding regions and likely encode nonfunctional proteins. Aberrant transcription of this gene may be involved in carcinogenesis of the lung, esophagus, and kidney.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLEC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DLEC1
Gene Accession No. Q9Y238
Gene Alias CFAP81; cilia and flagella associated protein 81; D630005C06Rik; deleted in lung and esophageal cancer 1; deleted in lung and esophageal cancer protein 1; deleted in lung and esophageal cancer protein 1 homolog; Deleted in lung cancer protein 1; DLC1; DLC-1; DLEC1; F56
Gene Symbols DLEC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DLEC1(Accession Q9Y238), corresponding to amino acid residues P308-Q358.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9940
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.