Learn More
R&D Systems™ DLin-KC2-DMA
SDP
Catalog No. 889510 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
88-951-0 10 mg
1 options
Catalog No. 88-951-0 Supplier R&D Systems™ Supplier No. 8895/10
Customers require a valid business account associated to their profile to access pricing and ordering options.
A Tocris product, part of R&D Systems. Ionizable cationic lipid
DLin-KC2-DMA is an ionizable cationic lipid (pKa = 6.7), used in the formulation of lipid nanoparticles (LNPs) encapsulating siRNA or plasmid DNA for non-toxic delivery into cells in vitro and in vivo.
For more information on LNPs and available components see our Lipid Nanoparticles page .
Specifications
Content And Storage Store at -20°C
Product Line Tocris
Quantity 10 mg