Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dilinoleyl methanol | 1169768-28-8 | MFCD32062843 | 98.0% | C37H68O | 10MG
SDP

Supplier:  Medchemexpress LLC HY14298610MG

Encompass_Preferred

Dilinoleyl methanol is an ionizable, unsaturated long-chain alcohol used as a lipid component and synthetic intermediate in lipid-based drug delivery systems and in the synthesis of ionizable cationic lipids. It is a liquid lipid (C37H68O, MW 528.94) commonly used in formulation development for lipid nanoparticles.

  • Ionizable lipid suitable for lipid nanoparticle formulations.
  • Useful synthetic intermediate for ionizable cationic lipid synthesis.
  • High purity (98.0%).
  • Liquid, colorless to light yellow; density ≈0.864 g/cm3.
  • Recommended storage: -20°C for long term; in solvent store at -80°C.

Catalog No. 50-002-58971


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.