Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572199
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572199 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572199 Supplier Invitrogen™ Supplier No. PA572199
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DLK1 Polyclonal Antibody for Western Blot, IHC (P), Flow

The Delta-like 1 homolog (DLK1) is a transmembrane protein containing six epidermal growth factor repeats. It is involved in the differentiation of several cell types, including adipocytes and is also thought to be a tumor suppressor. The DLK1 gene is one of several imprinted genes located in a region on chromosome 14q32; certain mutations in this imprinted region can cause phenotypes similar to maternal and paternal uniparental disomy of chromosome 14 (UPD14). DLK1 is expressed from the paternal allele; a polymorphism within this gene has been associated with child and adolescent obesity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLK1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% ProClin 300; pH 7.4
Gene DLK1
Gene Accession No. P80370, Q09163
Gene Alias Adipocyte differentiation inhibitor protein; AW742678; delta like non-canonical Notch ligand 1; Delta1; delta-like 1 homolog; delta-like 1 homolog (Drosophila); DLK; Dlk1; DLK-1; DlkI; FA1; Fetal antigen 1; Ly107; Peg9; pG2; Preadipocyte factor 1; preadipocyte factor-1; PREF; Pref1; pref-1; Protein delta homolog 1; SCP1; Scp-1; secredeltin; stromal cell derived protein 1; ZOG
Gene Symbols DLK1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 368-402 amino acids from the C-terminal region of human DLK1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13386, 8788
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.