Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLL1 (delta-like 1) Monoclonal Antibody (HMD1-5), Functional Grade, eBioscience™
GREENER_CHOICE

Catalog No. 5014569
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-145-69 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-145-69 Supplier Invitrogen™ Supplier No. 16576785
Only null left
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Description: The HMD1-5 monoclonal antibody reacts with mouse Delta-like 1, one of five type I transmembrane proteins that serves as a Notch receptor ligand. Upon binding the Notch receptor (e.g. Notch 1-4), Delta-like 1 undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases an intracellular fragment whose biological function remains controversial. Delta-like 1 is expressed by thymic stromal cells, as well as on macrophages, dendritic cells, and stromal cells in the spleen. This protein is also expressed in nonhematopoietic tissues such as the lung and brain. Delta-like 1 has been linked to B cell development and differentiation, especially marginal zone and plasma cells, regulation of splenic dendritic cells, and leukemogenesis. This monoclonal antibody has been reported to have neutralizing activity. Applications Reported: This HMD1-5 antibody has been reported for use in functional assays. Applications Tested: This HMD1-5 antibody has been tested by flow cytometric analysis of Delta-like 1-transfected CHO cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and ha...

Delta-like protein 1 (DLL1) is a transmembrane protein characterized by epidermal growth factor (EGF)-like repeats. It is a ligand for Notch receptor. It blocks the differentiation of progenitor cells into the B-cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLL1 (delta-like 1)
Applications Functional Assay
Classification Monoclonal
Clone HMD1-5
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene DLL1
Gene Accession No. Q61483
Gene Alias C-Delta-1; Delta; delta like canonical Notch ligand 1; delta like-1; DELTA1; delta-like 1; delta-like 1 (Drosophila); delta-like protein 1; DL1; Dll1; Dll1-derived cell-associated form; Dll1-EC; Dll1-IC; Dll1-intracellular form; Dll1-soluble form; Dll1-TMIC; Drosophila Delta homolog 1; H-Delta-1; LOW QUALITY PROTEIN: delta-like protein 1; OTTHUMP00000017690; UNQ146/PRO172
Gene Symbols DLL1
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13388
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.