Learn More
Echelon Biosciences Research Labs DLPG 250 mg
SDP
Supplier:  Echelon Biosciences Research Labs L5112250MG
Encompass
1,2-Dilauroyl-sn-glycero-3-phosphoglycerol, sodium salt (DLPG) is a form of phosphatidylglycerol (PG), a minor component (1-2%) of most of cell membranes. It can be found at higher concentrations (up to 11% of the total lipid content) in lung surfactant. It s presence in amniotic fluid indicates fetal lung maturity and is the basis of clinical testing for fetal fitness. PG is also used by cells as an intermediate in cardiolipin synthesis. Bacterial membranes contain up to 20% of PG. This item is non returnable
Catalog No. 50-720-514
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.