Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLX2 Recombinant Rabbit Monoclonal Antibody (HL1942)
GREENER_CHOICE

Catalog No. PIMA547229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547229 Supplier Invitrogen™ Supplier No. MA547229
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. This gene is located in a tail-to-tail configuration with another member of the gene family on the long arm of chromosome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLX2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1942
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DLX2
Gene Accession No. Q07687, Q64204
Gene Alias AW121999; DII A; distal-less homeo box 2; distal-less homeobox 2; DLX2; Dlx-2; Dlx5; DLX-5; homeobox Dlx5; homeobox protein DLX-2; Homeobox protein TES-1; TES1; Tes-1
Gene Symbols DLX2
Host Species Rabbit
Immunogen Recombinant fragment of human DLX2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1746, 296499
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.