Learn More
Medchemexpress LLC DM1-SMe | 138148-68-2 | 99.1% | 784.38 g/mol | C36H50ClN3O10S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1001285MG
Encompass_Preferred
DM1-SMe is an unconjugated maytansinoid cytotoxin that functions as a potent microtubule inhibitor and is used as a research reagent for studying antibody-drug conjugate (ADC) payload behavior. It is supplied as a solid and in solution for stock preparation, and is characterized with a defined molecular formula and high purity for laboratory applications.
  • Potent microtubule inhibitor for cytotoxicity studies.
  • Suitable as an ADC payload research reagent.
  • High purity supports analytical and biological testing.
  • Available in small-scale quantities and solution stocks for convenience.
  • Supplied with standard safety and technical documentation.
Catalog No. 50-002-15215
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.