Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DM4 Recombinant Rabbit Monoclonal Antibody (3H3)
GREENER_CHOICE

Catalog No. PIMA544657 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544657 Supplier Invitrogen™ Supplier No. MA544657
  • $576.00
    $504.65 / Each of 1

    Save $71.35

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Application notes: Indirect ELISA.

Ravtansine (DM4) is a maytansinoid, a chemical derivative of maytansine being investigated as the cytotoxic payload of a number of antibody-drug conjugates (ADCs). Microtubules are dynamic cytoskeletal polymers that switch stochastically between states of growing and shortening, called dynamic instability. They function in the precise segregation of chromosomes during cell division, transport of cellular cargos, and positioning and movement of intracellular organelles. Inhibition of microtubule function leads to cell cycle arrest and cell death. Microtubule-targeted drugs including the Vinca alkaloids, taxanes, and epothilones suppress the dynamic instability of microtubules, induce mitotic arrest, inhibit cell proliferation and induce apoptosis. The anticancer properties of maytansinoids have been attributed to their ability to disrupt microtubule function. The maytansinoid emtansine (DM1), for example, binds at the ends of microtubules and thereby suppress their dynamic instability. It is synthesized in order to link maytansinoids to antibodies via disulfide bonds. Maytansinoids inhibit tubulin polymerization and microtubule assembly and enhance microtubule destabilization, so there is potent suppression of microtubule dynamics resulting in a mitotic block and subsequent apoptotic cell death. DM4 can be used in the preparation of antibody drug conjugate. Although S-methyl DM1 and S-methyl DM4 inhibited microtubule assembly more weakly than maytansine, they suppressed dynamic instability more strongly than maytansine. Like vinblastine, the maytansinoids potently suppress microtubule dynamic instability by binding to a small number of high affinity sites, most likely at microtubule ends. Thus, the maytansine derivatives that result from cellular metabolism of the antibody conjugates are themselves potent microtubule poisons, interacting with microtubules as effectively as or more effectively than the parent molecule.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DM4
Applications ELISA
Classification Recombinant Monoclonal
Clone 3H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias Ravtansine
Host Species Rabbit
Immunogen DM4 coupled with OVA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.