Learn More
Cayman Chemical Indole-3-carbInol 5g
SDP
Supplier:  Cayman Chemical 113255 G
Encompass_Preferred
FK-506 s a potent mmunosuppressant that forms a hgh affnty complex (K 0 2 nM) wth FK-506 bndng proten 12 (FKBP12) Ths complex nhbts the actvty of the calcum/calmoduln-dependent proten phosphatase calcneurn leadng to dsrupton of T-cell actvaton 13842 SLF s a cell-permeable analog of FK-506 that bnds tghtly to FKBP but lacks the ablty to nhbt calcneurn 13706 By tself SLF does not exhbt a sgnfcant cellular response However homodmers of SLF or heterodmers wth another proten lgand provde a method to promote the dmerzaton of lgand-specfc fuson protens to nduce sgnalng events wthn a cellular envronment 1370113706 For example SLF monomers connected wth a 10-atom lnker actvates Fas sgnalng va Fas-FKBP fuson protens wth an EC50 of 6 nM 13701
Catalog No. 50-295-4013
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.