Learn More
United States Biological Corporation DMEM CA -FREE LOW GLUCOSE W L-
Catalog No. NC9532977 Shop All United States Biological Corporation Products
Encompass_Preferred
Dulbecco’s MEM is the most widely used modification of BME. It contains a 4-fold higher concentration of amino acids and vitamins. Non-essential amino acids and certain essential trace elements were added. The bicarbonate concentration was increased. The standard formula for DMEM is with 1000mg/ml glucose. DMEM was originally developed for the culture of mice embryonic cells. Today, it finds a broad application of serum free culture of normal and transformed mouse and chicken cells.
Catalog No. NC9532977 Supplier United States Biological Corporation Supplier No. D98001010L
Add to Cart
Add to Cart
Dulbecco’s MEM is the most widely used modification of BME. It contains a 4-fold higher concentration of amino acids and vitamins. Non-essential amino acids and certain essential trace elements were added. The bicarbonate concentration was increased. The standard formula for DMEM is with 1000mg/ml glucose. DMEM was originally developed for the culture of mice embryonic cells. Today, it finds a broad application of serum free culture of normal and transformed mouse and chicken cells.