Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech DMEM/F12 (1:1) Cell Culture Medium, w: stable Glutamine, w: 1.2 g/L NaHCO3 (10 x 500 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0441150X10
Encompass_Preferred
PAN-Biotech DMEM/F12 (1:1) Cell Culture Medium, w: Stable Glutamine, w: 1.2 g/L NaHCO3. QUANTITY: 10 x 500 ml. MANUFACTURED BY: PAN-Biotech GmbH (Germany). BICARBONATE: With 1.2 g/L Sodium Bicarbonate. GLUCOSE: with glucose. GLUTAMINE: with Stable Glutamine. HEPES: Without HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: with Sodium pyruvate. STERILE: Yes. STORAGE TEMPERATURE: +2-8°C. SUMMARY: DMEM/F12 Medium is a 1:1 mixture of Dulbecco’s Modified Essential Medium (DMEM) and Ham’s F-12 Medium, commonly used in growing a variety of mammalian cell cultures. DMEM/F12 is rich and complex, containing all 21 amino acids, 10 vitamins, glucose, zinc, and iron among other components. Together, it combines DMEM's high concentration of amino acids and vitamins of DMEM and the higher concentration of zinc sulfate, putrescine and linoleic acid of F12.
Catalog No. 50-238-8570
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.