Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech DMEM Cell Culture Medium, w: 4.5 g/L Glucose, w: L-Glutamine, w: Sodium pyruvate, w: 3.7 g/L NaHCO3 (500 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0403590
Encompass_Preferred
PAN-Biotech DMEM Cell Culture Medium, w: 4.5 g/L Glucose, w: L-Glutamine, w: Sodium pyruvate, w: 3.7 g/L NaHCO3. QUANTITY: 500 ml. MANUFACTURED BY: PAN-Biotech GmbH (Germany). BICARBONATE: With 3.7 g/L Sodium Bicarbonate. GLUCOSE: High Glucose (4.5 g/L). GLUTAMINE: with L-Glutamine. HEPES: Without HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: With Sodium Pyruvate. STERILE: Yes. STORAGE TEMPERATURE: +2°C - +8°C. SUMMARY: Intrinsically developed for the cultivation of murine embryonic cells, DMEM (Dulbecco's Modified Eagle Medium) is tailor-made for the cultivation of a broad range of cells, especially if the medium is supplemented with FBS. DMEM is an Eagle medium modification with a fourfold content of amino acids and vitamins. DMEM with 1.0 g/L Glucose is the standard medium, whereas DMEM with 4.5 g/L Glucose is for cells which have a high energy demand.
Catalog No. 50-238-8636
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.