Learn More
Apexbio Technology LLC Berberine(Synonyms: Berberine Hydrochloride, Umbellatine, Berberin, Natural Yellow 18), 20mg, CAS: 2086-83-1.
SDP
Supplier:  Apexbio Technology LLC N136820
Encompass_Preferred
Berberine (CAS 2086-83-1) is an isoquinoline alkaloid isolated primarily from Cortex Phellodendri Chinensis It exhibits diverse pharmacological activities mediated predominantly through AMP-activated protein kinase (AMPK) activation and modulation of metabolic signaling pathways Berberine has been extensively studied for its effects on glucose and lipid metabolism inflammation regulation and antimicrobial activity Researchers commonly employ berberine in metabolic disease models examining therapeutic potential in diabetes obesity and cardiovascular research Store dry sealed and protected from moisture and heat
Catalog No. 50-000-61378
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.