Learn More
Apexbio Technology LLC DMG-PEG2000-NH₂(Synonyms: DMG-PEG2000-Amine, DMG-PEG2K-NH₂, Dimyristoyl glycerol PEG2000 amine, DMG-PEG-Amine), 100mg.
SDP
Supplier:  Apexbio Technology LLC M2006100
Encompass_Preferred
DMG-PEG2000-NH is a polyethylene glycol (PEG) derivative functionalized with a primary amine (-NH ) group It readily participates in amide bond formation reactions with carboxyl-containing biomolecules such as proteins peptides or other compounds bearing compatible functional groups Due to this reactive amine terminus DMG-PEG2000-NH serves as a versatile linker for constructing lipid-based drug delivery platforms including liposomes and lipid nanoparticles (LNPs) specifically for encapsulating therapeutic agents like siRNA Its application facilitates conjugation or modification of biomolecules enhancing stability solubility and biocompatibility
Catalog No. 50-000-61343