Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529110 Supplier Invitrogen™ Supplier No. PA529110
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG conditioned medium. Predicted reactivity: Mouse (97%), Rat (97%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Calcium-binding protein which may play a role in dentin mineralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMP4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene FAM20C
Gene Accession No. Q8IXL6
Gene Alias C76981; Dentin matrix protein 4; DMP4; DMP-4; Extracellular serine/threonine protein kinase FAM20C; Fam20c; family with sequence similarity 20 member C; family with sequence similarity 20, member C; GEF-CK; Golgi casein kinase; golgi-enriched fraction casein kinase; Kiaa4081; mKIAA4081; RGD1311980; RNS
Gene Symbols FAM20C
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DMP4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56975
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.