Learn More
Echelon Biosciences Research Labs DMPE 1 g
SDP
Supplier:  Echelon Biosciences Research Labs L21141G
Encompass
1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine (DMPE) is a water soluble derivative of phosphatidylethanolamine with (14:0) myristic acid acyl chains. Phosphatidylethanolamine (PE) is formed primarily in the reaction of CDP-ethanolamine and diacylglycerol. PE is one of the major components of cell membranes in bacteria and in nervous system. It is useful in liposome production. This item is non returnable
Catalog No. 50-720-500
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.