Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMRT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585040
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585040 Supplier Invitrogen™ Supplier No. PA585040
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNGYGSPYLY MGGPVSQPPR APLQRTPKC Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

Probable transcription factor that plays a role in configuring the spinal circuits controlling stride in vertebrates. Involved in neuronal specification within specific subdivision of spinal cord neurons and in the development of a coordinated locomotor network controlling limb movements. May regulate transcription during sexual development. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRT3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DMRT3
Gene Accession No. Q9NQL9
Gene Alias DMRT3; DMRTA3; DMRT-like family A3; Doublesex and mab-3 related transcription factor 3; doublesex- and mab-3-related transcription factor 3; testis-specific protein
Gene Symbols DMRT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DMRT3. Recombinant protein control fragment (Product #RP-108690).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58524
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.