Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMRTA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560821 Supplier Invitrogen™ Supplier No. PA560821
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSNKPDSILS PHPGEQSGGE ESPRSLSSSD LESGNESEWV KDLTATKASL PTVSSRPRDP LDILTKIFPN YRRSRLEGIL RFCK Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 75%.

DMRTA1 (DMRT Like Family A1) is a Protein Coding gene. Diseases associated with DMRTA1 include Bartholin's Duct Cyst and Cardiomyopathy, Dilated, 1J. Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity and sequence-specific DNA binding. An important paralog of this gene is DMRTA2. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRTA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DMRTA1
Gene Accession No. Q5VZB9
Gene Alias DMO; DMRT like family A1; DMRT4; Dmrta1; DMRT-like family A1; doublesex and mab-3 related transcription factor 4; doublesex and mab-3 related transcription factor like family A1; doublesex- and mab-3-related transcription factor 4; Doublesex- and mab-3-related transcription factor A1
Gene Symbols DMRTA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DMRTA1. Recombinant protein control fragment (Product #RP-99927).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63951
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.