Learn More
Cytiva (Formerly Pall Lab) DMSO-Safe Acrodisc Syringe Filter - 0.2 m, 25 mm, sterile (50/pkg)
Catalog No. NC1034197 Shop All Cytiva Products
Encompass
DMSO-Safe Acrodisc syringe filters feature a polypropylene housing and nylon membrane that are compatible with DMSO.They have excellent chemical compatibility with esters, bases, and alcohols. They are ideal for the sterilization of media used for cell cryopreservation.The DMSO-Safe Acrodisc syringe filters are sterilized by ethylene oxide and individually packaged. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1034197 Supplier Cytiva Supplier No. 4433
DMSO-Safe Acrodisc syringe filters feature a polypropylene housing and nylon membrane that are compatible with DMSO.They have excellent chemical compatibility with esters, bases, and alcohols. They are ideal for the sterilization of media used for cell cryopreservation.The DMSO-Safe Acrodisc syringe filters are sterilized by ethylene oxide and individually packaged. This product is not approved for use in medical, clinical, surgical or other patient applications.