Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DMSO, sterile filtered
GREENER_CHOICE

Catalog No. 31765ML
Change view
Click to view available options
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
31-765-ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 31-765-ML Supplier Tocris Bioscience™ Supplier No. 3176/5ML
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Solvent for polar and nonpolar compounds

DMSO, sterile filtered is a solvent with wide ranging applications in cell culture, cryopreservation and organic chemistry. DMSO dissolves both polar and nonpolar compounds. Improves responsiveness of human embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) to differentiation signals. DMSO is used in PCR to inhibit secondary structure formation by DNA template or primers.

Specifications

Chemical Name or Material Dimethyl sulfoxide
CAS 67-68-5
Synonym Methyl sulfoxide
Quantity 5 mL
Target Buffers, Chelators and Reagents
Molecular Formula C2H6OS
Formula Weight 78.13
Purity 99.9%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.