Learn More
Sigma Aldrich Fine Chemicals Biosciences DMT-2'Fluoro-dU Phosphoramidite configured for ABI | 146954-75-8 | MFCD11113919 | 1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences U2112301G
Encompass
DMT-2'Fluoro-dU Phosphoramidite configured for ABI | Purity: >=99% (31P-NMR); >=99.0% (reversed phase HPLC); conforms (1H-NMR); conforms (LC/MS) | Mol Wt: 748.78 | 146954-75-8 | MFCD11113919 | 1G
Catalog No. 50-174-2639
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.