Learn More
Description
DNA Adenine Methylase Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | DNA Adenine Methylase |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | RB1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene Alias | DAM; DAM methylase |
| Host Species | Mouse |
| Immunogen | Recombinant DNA Adenine Methylase protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
