Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA Damage Competitive ELISA Kit

Catalog No. EIADNADX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIADNADX10 10 x 96 Tests
EIADNAD 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIADNADX10 Supplier Invitrogen™ Supplier No. EIADNADX10
Add to Cart
Add to Cart
Request a Quote

ELISA

The DNA Damage ELISA quantitates PGFM in serum, plasma, saliva, urine, digested DNA, fecal extracts, or cell culture medium. The assay will exclusively recognize both natural and recombinant DNA Damage. Principle of the method The PGFM solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is inversely proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Free radicals and other reactive species are constantly generated in vivo and cause oxidative damage to biomolecules, a process held in check only by the existence of multiple antioxidant and repair systems as well as the replacement of damaged nucleic acids, proteins, and lipids. Intracellular free radical species (ROS) are produced as a result of normal metabolism and extracellular forms are produced as a result of ultraviolet radiation or ionizing radiation. Cellular function may be interrupted or stopped if DNA damage corrupts the integrity of essential information contained in the genome. When individual bases are damaged, nonspecific DNA repair enzymes excise DNA lesions to release deoxynucleotides, and base specific repair glycosylases excise the corresponding base. Deoxynucleotides are enzymatically hydrolyzed to stable deoxynucleosides, and these repair products are transported through the blood and excreted in the urine. Damage to RNA is reflected in nucleoside adducts. Among numerous types of oxidative DNA damage, the formation of 8-hydroxy-2'-deoxyguanosine (8-OHdG) is a ubiquitous marker of oxidative stress. 8-OHdG is physiologically formed and enhanced by chemical carcinogens. During the repair of damaged DNA in vivo by exonucleases, the resulting 8-OHdG is excreted without further metabolism into urine. 8-Hydroxy-2'-deoxyguanosine (8-OHdG) is identical across all species.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 62.6-8000 pg/mL
Assay Sensitivity 50.9 pg/mL
Description DNA Damage Competitive ELISA Kit
Assay Multiplex Assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Digested DNA, Fecal Extract, Plasma, Saliva, Serum, Supernatant, Urine
Sufficient For 10 x 96 Assays
Target Species Chemical
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Absorbance Microplate Reader
Interassay CV 9.9%
Intraassay CV 9%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Research Discipline Inflammation
Sample Volume Digested DNA 50 μL, Fecal Extract 50 μL, Plasma 6.25 μL, Saliva 25 μL, Serum 6.25 μL, Supernatant 50 μL, Urine 12.5 μL
Shipping Condition Wet or Dry Ice
Storage Requirements -20°C
Target DNA Damage
Test Time 1 hr 20 min
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.