Learn More
Edvotek™ DNA DuraGel™
SDPSCI_ED
Catalog No. S05711 Shop All EDVOTEK Products
Change view
Click to view available options
Device Holds:
6 Gels (12 to 24 Students)
6 Gels (4 to 8 Students)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Device Holds
S05711 6 Gels (12 to 24 Students)
S05712 6 Gels (4 to 8 Students)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S05711 Supplier Edvotek™ Supplier No. S43
Add to Cart
Add to Cart
Designed for students to learn pipetting samples in agarose gels.
  • DNA DuraGel™ gels are permanent polymer gels that allow students to practice the critically important skill of pipetting/gel loading.
  • The clear, reusable gels are designed for the practice of loading 5 - 35 μl of samples.
  • The gel grids are imprinted with a ruler for sizing DNA fragments.Also included are simulated FlashBlue™ and Ethidium Bromide gel images, ideal for representing how actual gels are stained with Methylene Blue and Ethidium Bromide.
Specifications
Device Holds 6 Gels (12 to 24 Students)
Type DNA DuraGel™
Includes 6 Reusable DNA DuraGel™, 4 FlashBlue™ and 4 Ethidium Bromide gel images, practice gel loading solution and mini-transfer pipets