Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies DNA Manipulatives
SDPSCI_ED

Catalog No. S65112 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S65112 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65112 Supplier United Scientific Supplies Supplier No. DNAKIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Magnetic teacher manipulatives designed to be used on the chalk board
  • Student manipulative sets for individual study
  • Experiments using laminated teacher and student gel electrophoresis boards
  • Instructional CD-ROM

The teacher-developed DNA manipulatives kit is perfect for helping students to understand recombinant DNA technology.

  • Combines real-world applications with instructor demonstrations and hands-on student activities
  • Introduces the structure and function of DNA (chromosomal and plasmid)
  • Allows students to “insert” antibiotic-resistant genes into plasmids
  • Manipulative plasmids include information on the number of base pairs and lessons about restriction enzymes
  • Teacher board is large enough to be seen by the class for demonstrations
  • Students use a set of 15 boards

Specifications

Grade 9 to 12
For Use With (Application) For helping students to understand recombinant dna technology
Includes Magnetic teacher manipulatives, student manipulative sets, experiments using laminated teacher and student gel electrophoresis boards, instructional cd-rom
Class Size 30 Students
Product Type DNA Manipulative Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.