Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DNA Marker, high range, 1,000 Base Pair Ladder

Catalog No. AAJ63178S0
Change view
Click to view available options
Quantity:
1 mL
2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63178S0 2 mL
AAJ63178XF 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63178S0 Supplier Thermo Scientific Chemicals Supplier No. J63178S0
Add to Cart

DNA ladders are used in gel electrophoresis to determine the size and quantity of testing DNA fragments of genomic, plasmid, and PCR DNA. It is commonly used in the studies of plasmid DNA and PCR products. 1 kb DNA Ladder has been used as a molecular marker to determine the molecular weight and size of double stranded DNA during electrophoresis. 1 kb Ladder contains 11 fragments consisting of 500 bp repeats from 0.5 to 3 kb, 1 kb repeats from 3 to 6 kb, and 2 kb repeats from 6 to 10 kb. In NA electrophoresis, five ?l of the marker should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel. Suitable for use in Northern and Southern blotting. All DNA markers can be stained with ethidium bromide, SYBR™ Green, and Nancy-520.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Quantity 2 mL
Storage Requirements −30° to −10°C
Product Type Molecular Weight Marker

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.