Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DNA Methyltransferase Inhibitor

Catalog No. AAJ64630MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64630MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64630MA Supplier Thermo Scientific Chemicals Supplier No. J64630MA
Get 15% Off Thermo Scientific Reagents
Use promo code REAGENTS15
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 48208-26-0 | C19H14N2O4, C19H14N2O4 | 334.33 g/mol

A cell-permeable, specific DNA methyltransferases inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 48208-26-0
Molecular Formula C19H14N2O4
Synonym RG108; N-Phthalyl-L-tryptophan

Specifications

Quantity 10 mg
Description Yellow
For Use With (Application) A cell-permeable, specific DNA methyltransferases inhibitor
Formula Weight 334.32g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Solid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.