Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Monoclonal Antibody (ET844.3.1), Invitrogen™

Mouse Monoclonal Antibody
$472.00 - $1160.00
Specifications
| Antigen | DNA |
|---|---|
| Clone | ET844.3.1 |
| Concentration | 0.8 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA546039
|
Invitrogen™
MA546039 |
100 μg |
Each for $472.00
|
|
|||||
01-671-554
![]() |
Invitrogen™
MA110601 |
500 μg |
Each for $671.00
|
|
|||||
|
PIMA545999
|
Invitrogen™
MA545999 |
1 mg |
Each for $1,160.00
|
|
|||||
Description
This antibody recognizes single- or double-stranded eucaryotic DNA as well as E. coli DNA. Reactivity with other prokaryotic DNA has not been investigated.
DNA Monoclonal antibody specifically detects DNA in Many samples. It is validated for ELISA.Specifications
| DNA | |
| 0.8 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| Calf thymus DNA. | |
| Primary | |
| 4°C, do not freeze |
| ET844.3.1 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Chemical | |
| dsdna; e. coli dna; single-stranded DNA | |
| IgM | |
| PEG precipitation | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title