Learn More
Invitrogen™ DNA-PK Polyclonal Antibody

Description
Heat-mediated antigen retrieval using an antigen unmasking fluid is recommended, followed by a 2-hr incubation with primary antibody. Rabbit anti DNA PKcs antibody recognizes the human DNA-dependent protein kinase catalytic subunit, also known as DNA-PKcs, DNPK1 or p460.
Specifications
Specifications
| Antigen | DNA-PK |
| Applications | ChIP Assay, Gel Shift, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with <0.1% sodium azide |
| Gene | PRKDC |
| Gene Accession No. | P78527, P97313 |
| Gene Alias | AI326420; AU019811; DNA-dependent protein kinase catalytic subunit; DNAPDcs; DNAPK; DNA-PK catalytic subunit; DNA-PKcs; DNPK1; DOXNPH; doxorubicin nephropathy; dxnph; hyper-radiosensitivity of murine scid mutation, complementing 1; HYRC; HYRC1; I79_017613; IMD26; p350; p460; Prkdc; protein kinase, DNA activated, catalytic polypeptide; protein kinase, DNA-activated, catalytic polypeptide; scid; severe combined immunodeficiency; slip; Xrcc7 |
| Gene Symbols | PRKDC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.