Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase I, 2500 U, G-Biosciences
SCI_ED

Catalog No. S001154
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001154 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001154 Supplier G-Biosciences Supplier No. 7862265
Add to Cart
Add to Cart

DNA Polymerase I (E. coli) is a template-dependent DNA polymerase with intrinsic proofreading exonuclease (3' → 5') and nick translation exonuclease (5' → 3') activities.

  • DNA polymerase I plays a crucial role in prokaryotic DNA replication and repair by synthesizing DNA, proofreading for errors, and removing RNA primers.

Specifications

Concentration 10 U/μL
Detection Method Gel Electrophoresis
Form Liquid
GC-Rich PCR Performance Medium
PCR Method End-point PCR
Reaction Speed Standard
Label or Dye Fluorescein (FITC), FAM, Biotin
Product Type DNA Polymerase I
For Use With (Application) Isothermal Amplification, Microbial Detection, Pathogen Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.