Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase Kappa Antibody [Allophycocyanin], R&D Systems™
SDP

Catalog No. NB802237
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB802237 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB802237 Supplier R&D Systems Supplier No. NBP106559APC

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

DNA Polymerase Kappa Polyclonal antibody specifically detects DNA Polymerase Kappa in Human samples. It is validated for Immunoprecipitation,Western Blot

Specifications

Antigen DNA Polymerase Kappa
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias DINB protein, DINPPOLQDINB1EC 2.7.7.7, DNA polymerase kappa, polymerase (DNA directed) kappa
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 825 and 870 of human DNA polymerase kappa using the numbering given in entry NP_057302.1
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51426
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.