Learn More
Zymo Research Corporation Sample DNA/RNA Shield Fecal Collection Tube (with Beads, 10 Pack)
SDP
Supplier:  Zymo Research Corporation R1137FS
Encompass
Free Sample DNA/RNA Shield Fecal Collection tubes are designed for the collection and preservation of nucleic acids from stool specimens. Thesefecal collection tubes take a microbial snapshot of a sample while inactivating viruses making samples safe and ready for transport. Samples stored in these fecal collection tubes are stable at ambient temperature, and can be frozen for longer-term storage. Simply collect a spoonful of the specimen into the stool collection tubes prefilled with preservative and shake vigorously to ensure proper stabilization. R1137 is pre-filled with 5 glass beads to help facilitate homogenization.
Catalog No. NC3346005
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.