Learn More
Description
Specifications
Specifications
| Antigen | DNA-RNA Hybrid |
| Applications | ChIP Assay, Dot Blot, Immunocytochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | S9.6 |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Formulation | Purified mouse IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide. |
| Host Species | Mouse |
| Immunogen | DNA-RNA heteropolymer duplex prepared by transcription of phi X174 single-stranded DNA with DNA-dependent RNA polymerase (Boguslawski, S.J., et al. (1986). J. Immunol Methods. 89(1):123-130). |
| Purification Method | Protein G Purified |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.