Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA/RNA Oxidative Damage Markers Monoclonal Antibody (15A3)
GREENER_CHOICE

Catalog No. 01671555 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-555 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-555 Supplier Invitrogen™ Supplier No. MA110602
  • $663.00
    $506.65 / Each of 1

    Save $156.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-10602 detects DNA and RNA Oxidative Damage Markers from oxidatively damaged eucaryotic DNA samples. MA1-10602 has been successfully used in ELISA and immunohistochemistry applications. The MA1-10602 immunogen is 8-hydroxyguanosine, conjugated to BSA and casein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA/RNA Oxidative Damage Markers
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 15A3
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.1% sodium azide; pH 7.4
Gene Alias DNA/RNA Damage
Host Species Mouse
Immunogen 8-hydroxyguanosine, conjugated to BSA and casein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Multi-species
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.