Learn More
Zymo Research Corporation DNA/RNA Shield Fecal Collection Tube (10 pack)
Supplier: Zymo Research Corporation R1101
DNA/RNA Shield Fecal Collection tubes are designed for the collection and preservation of nucleic acids from stool specimens. These stool collection tubes take a microbial snapshot of a sample while inactivating viruses making samples safe and ready for transport. Samples stored in the these stool collection tubes are stable at ambient temperature, and can be frozen for longer-term storage. Simply collect a spoonful of the specimen into the stool collection tubes prefilled with preservative and shake vigorously to ensure proper stabilization.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.