Learn More
AB Sciex LLC DNA Size Standard Kit 400 bp
Supplier:  AB Sciex LLC 608098
Encompass
For analysis of fragments up to 400 nucleotides. Includes mineral oil. Sufficient for 96 fragment analysis separations. Includes DNA fragments of the following sizes labeled with CEQ WellRED fluorescent dye: 60, 70, 80, 90, 100, 120, 140, 160, 180, 190, 200, 220, 240, 260, 280, 300, 320, 340, 360, 380, 400, and 420 nucleotides. Storage temperature: -20C.
Catalog No. NC1661524
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.