Learn More
Enzo Life Sciences DNA topoisomerase II alpha monoclonal antibody (1C5) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC210E
Encompass_Preferred
Clone: 1C5. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human DNA-topoisomerase IIα (C-terminal fragment). UniProt ID: P11388. Species reactivity: Human. Applications: Flow Cytometry, ICC, IHC, IP, WB. Recommended Dilutions: Flow Cytometry (0.1-1.0µg/ml) Immunocytochemistry (1-10µg/ml) Immunohistochemistry (1-5µg/ml) Immunoprecipitation (1-10µg/ml, 200-300ml cell extract) Western Blot (0.1-0.5µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7244
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.