Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAH11 Monoclonal Antibody (C5), Invitrogen™

Catalog No. p-7240948
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543854 20 μL
PIMA543853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543854 Supplier Invitrogen™ Supplier No. MA543854
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This gene encodes a ciliary outer dynein arm protein and is a member of the dynein heavy chain family. It is a microtubule-dependent motor ATPase and has been reported to be involved in the movement of respiratory cilia. Mutations in this gene have been implicated in causing Kartagener Syndrome (a combination of situs inversus totalis and Primary Ciliary Dyskinesia (PCD), also called Immotile Cilia Syndrome 1 (ICS1)) and male sterility. [provided by RefSeq, Mar 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAH11
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene DNAH11
Gene Accession No. Q96DT5
Gene Alias axonemal beta dynein heavy chain 11; axonemal dynein heavy chain 11; CILD7; ciliary dynein heavy chain 11; DNAH11; DNAHBL; DNAHC11; DNHBL; DPL11; dynein axonemal heavy chain 11; dynein heavy chain 11, axonemal; dynein, axonemal, heavy chain 11; dynein, axonemal, heavy polypeptide 11; dynein, ciliary, heavy chain 11
Gene Symbols DNAH11
Host Species Mouse
Immunogen Leu3896-Asn4122 (Accession Q96DT5), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8701
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.