Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJB11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552746
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552746 Supplier Invitrogen™ Supplier No. PA552746
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPGVRDGMEY PFIGEGEPHV DGEPGDLRFR IKVVKHPIFE RRGDDLYTNV TISLVESLVG FEMDITHLDG HKVHISRDKI TRPGAKLWKK GEGLPNFDNN NIKGSLIITF DVDFPKEQLT EEAREGIKQL LKQGSVQKVY NGLQ Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

DNAJB11 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity bystimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus a glycine/phenylalanine (G/F)-rich region and a C-terminal cysteine-rich region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJB11
Gene Accession No. Q6TUG0, Q99KV1, Q9UBS4
Gene Alias 1810031F23Rik; ABBP2; ABBP-2; AL024055; APOBEC1-binding protein 2; cb954; Dj9; Dj-9; DnaJ (Hsp40) homolog, subfamily B, member 11; DnaJ heat shock protein family (Hsp40) member B11; dnaJ homolog subfamily B member 11; DnaJ protein 9; DnaJ protein homolog 9; DNAJB11; dnajb11 protein; DnaJ-like protein subfamily B member 11-like protein; EDJ; Endoplasmic reticulum DNA J domain-containing protein 3; ER-associated DNAJ; ER-associated DNAJ protein 3; ER-associated Hsp40 co-chaperone; ERdj3; ERJ3; ERj3p; ER-resident protein ERdj3; HDJ9; hDj-9; HEDJ; Human DnaJ protein 9; Liver regeneration-related protein LRRGT00084; LRRGT00084; PRO1080; PSEC0121; PWP1-interacting protein 4; UNQ537; UNQ537/PRO1080
Gene Symbols DNAJB11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAJB11. Recombinant protein control fragment (Product #RP-90018).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360734, 51726, 67838
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.