Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJB12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589947 Supplier Invitrogen™ Supplier No. PA589947
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVHVYSNGRM RYTYQQRQDR RDNQGDGGLG VFVQLMPILI LILVSALSQL MVSSPPYSLS PRPSVGHIHR RVTDHLGVVY YVGDTFSEEY TGSSLKTVER NVEDDYIANL RNNCWKEKQQ KEGLLYRARY FGDTDMYHRA QKMGTPSCSR LSEVQASLHG; Positive Samples: 293T, HT-29, BxPC-3, Mouse kidney, Mouse liver, Rat spleen; Cellular Location: Membrane, Single-pass membrane protein.

DNAJB12 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DNAJB12
Gene Accession No. Q9NXW2, Q9QYI4
Gene Alias DJ10; DKFZp586B2023; DnaJ (Hsp40) homolog, subfamily B, member 12; DnaJ heat shock protein family (Hsp40) member B12; DnaJ homolog 10; dnaJ homolog subfamily B member 12; DnaJ subfamily B member 12-like protein; Dnajb12; LOW QUALITY PROTEIN: dnaJ homolog subfamily B member 12; mDj10
Gene Symbols DNAJB12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 250-409 of human DNAJB12 (NP_0600963).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294513, 54788, 56709
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.