Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJB4 Monoclonal Antibody (OTI2F8), TrueMAB™, OriGene
SDP

Catalog No. 501684429
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4429 Supplier OriGene Supplier No. TA812139
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DnaJ heat shock induced proteins are from the bacterium Escherichia coli and are under the control of the htpR regulatory protein. The DnaJ proteins play a critical role in the HSP 70 chaperone machine by interacting with HSP 70 to stimulate ATP hydrolysis. The proteins contain cysteine rich regions that are composed of zinc fingers that form a peptide binding domain responsible for the chaperone function. DnaJ proteins are important mediators of proteolysis and are involved in the regulation of protein degradation, exocytosis and endocytosis. DnaJB4 (DnaJ homolog subfamily B member 4), also known as HLJ1, is expressed in skeletal muscle, heart and pancreas, and lower expression in brain, placenta and liver.

Specifications

Antigen DNAJB4
Applications Western Blot
Classification Monoclonal
Clone OTI2F8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DNAJB4
Gene Accession No. Q9UDY4
Gene Alias DNAJW, DjB4, HLJ1
Gene Symbols DNAJB4
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 2-337 of human DNAJB4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11080
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.