Learn More
DNAJC13 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304872AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
DNAJC13, also known as receptor-mediated endocytosis 8 (RME8), is the human homolog to a DnaJ domain-containing protein originally identified in a screen for endocytic defects in C. elegans. It is thought to be a co-chaperone of Hsc70 which regulates protein conformation at membrane sites and plays a role in intracellular trafficking, co-localizing with markers of the endosomal system. Recent experiments have indicated that the DNAJC13 protein is involved in membrane trafficking through early endosomes but not through degradative organelles. DNAJC13 has been also been shown to regulate the intracellular trafficking of the epidermal growth factor receptor.Specifications
DNAJC13 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O75165 | |
Dnajc13 | |
Between 1175 and 1225 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DNAJC13 | |
DNAJ homolog subfamily C member 13, receptor-mediated endocytosis 8, RME8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23317, 235567 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.