Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC15/MCJ Mouse anti-Human, KHL, Clone: WeNA12, MilliporeSigma™

Catalog No. MABS21482MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS21482MI 25 μL
MABS21481MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS21482MI Supplier MilliporeSigma Supplier No. MABS214825UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNAJC15/MCJ Monoclonal antibody specifically detects DNAJC15/MCJ in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC15/MCJ
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone WeNA12
Conjugate KLH
Gene Accession No. NP_037370
Gene Alias DnaJ homolog subfamily C member 15;Cell growth-inhibiting gene 22 protein;Methylation-controlled J protein;MCJ
Gene Symbols DNAJC15, DNAJD1, GIG22, HSD18
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to the first 35 amino acids from the N-terminal region of human Methylation-controlled J protein (DNAJC15).
Molecular Weight of Antigen 16.38 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone WeNA12 is a mouse monoclonal antibody that detects human Methylation-controlled J protein (DNAJC15). It targets an epitope within the first 35 amino acids from the mitochondrial intermembrane N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.