Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565250 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565250 Supplier Invitrogen™ Supplier No. PA565250
  • $617.00
    $498.65 / Each of 1

    Save $118.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: GETGFNSSCFQ EWAKLYEFVH SFQDEKRQLA YQVLGLSEGA TNEEIHRSYQ ELVKVWHPDH NLDQTEEAQR HFLEIQAAYE VLSQPRKP Highest antigen sequence identity to the following orthologs - mouse 84%, rat 85%.

May function as a co-chaperone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC22
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJC22
Gene Accession No. Q8N4W6
Gene Alias 2810451A06Rik; AI506245; DnaJ (Hsp40) homolog, subfamily B, member 3; heat shock protein, DNAJ-like 3; DnaJ (Hsp40) homolog, subfamily C, member 22; DnaJ heat shock protein family (Hsp40) member C22; dnaJ homolog subfamily C member 22; Dnajc22; RGD1311098; wurst homolog; wus
Gene Symbols DNAJC22
Host Species Rabbit
Immunogen Recombinant Human DNAJC22. Recombinant protein control fragment (Product #RP-104709).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79962
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.