Learn More
Invitrogen™ DNAJC28 Polyclonal Antibody

Description
PA1-9539 detects C21 in human samples. PA1-9539 has been successfully used in Western blot procedures. By Western blot, PA1-9539 detects a 40 kDa protein representing C21. PA1-9539 immunogen is a purified recombinant C21 corresponding to the chromosome 21 gene (Human).
Specifications
Specifications
| Antigen | DNAJC28 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | DNAJC28 |
| Gene Accession No. | Q9NX36 |
| Gene Alias | BC020175; C21; C21orf55; C21orf78; DC42; DnaJ (Hsp40) homolog, subfamily C, member 28; DnaJ heat shock protein family (Hsp40) member C28; dnaJ homolog subfamily C member 28; DNAJC28; FLJ12894; IRA1; ORF28; TBLR2 |
| Gene Symbols | DNAJC28 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.