Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAL4 Monoclonal Antibody (3C10A4)
GREENER_CHOICE

Catalog No. PIMA531720
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531720 Supplier Invitrogen™ Supplier No. MA531720
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31720 has been tested in indirect ELISA.

Force generating protein of respiratory cilia. Produces force towards the minus ends of microtubules. Dynein has ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAL4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3C10A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene DNAL4
Gene Accession No. O96015
Gene Alias D15Ertd424e; DNAL4; Dnalc4; dynein axonemal light chain 4; dynein light chain 4, axonemal; dynein light chain, outer arm 4; dynein, axon, light chain 4; dynein, axonemal, light chain 4; dynein, axonemal, light polypeptide 4; MRMV3; outer arm dynein light chain 4; PIG27; proliferation-inducing gene 27; proliferation-inducing protein 27
Gene Symbols DNAL4
Host Species Mouse
Immunogen Purified recombinant fragment of human DNAL4 (AA: 1-105) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10126
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.